missing translation for 'onlineSavingsMsg'
Learn More

Di-2-ethylhexylamine, 99%, Thermo Scientific Chemicals

Catalog Number 10043360
Click to view available options
Quantity:
250 mL
1 L
2.5 L
This item is not returnable. View return policy
106-20-7
C16H35N
241.46
MFCD00009489
SAIKULLUBZKPDA-UHFFFAOYSA-N
bis 2-ethylhexyl amine, di 2-ethylhexyl amine, diisooctylamine, 1-hexanamine, 2-ethyl-n-2-ethylhexyl, bis-2-ethylhexylamine, 2,2'-diethyldihexylamine, 2,2'-diethylhexylamine, di-2-ethylhexyl amine, dihexylamine, 2,2'-diethyl, ccris 4619
7791
2-ethyl-N-(2-ethylhexyl)hexan-1-amine
CCCCC(CC)CNCC(CC)CCCC
This item is not returnable. View return policy
106-20-7
C16H35N
241.46
MFCD00009489
SAIKULLUBZKPDA-UHFFFAOYSA-N
bis 2-ethylhexyl amine, di 2-ethylhexyl amine, diisooctylamine, 1-hexanamine, 2-ethyl-n-2-ethylhexyl, bis-2-ethylhexylamine, 2,2'-diethyldihexylamine, 2,2'-diethylhexylamine, di-2-ethylhexyl amine, dihexylamine, 2,2'-diethyl, ccris 4619
7791
2-ethyl-N-(2-ethylhexyl)hexan-1-amine
CCCCC(CC)CNCC(CC)CCCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -60.0°C
Color Colorless to Yellow
Density 0.8000g/mL
Boiling Point 281.0°C
Flash Point 128°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3(CH2)3CH(C2H5)CH2)2NH
Refractive Index 1.4415 to 1.4435
Quantity 250 mL
Beilstein 04,388
Specific Gravity 0.8
Solubility Information Solubility in water: <0.2g/L (20 c) in water. Other solubilities: soluble in hydrocarbons
Viscosity 3.7 mPa.s (20°C)
Formula Weight 241.46
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Di-2-ethylhexylamine, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Di-2-ethylhexylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'