missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DHX9 Recombinant Rabbit Monoclonal Antibody (24GB4140)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557014
This item is not returnable.
View return policy
Description
DHX9 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The human alanyl-tRNA synthetase (AARS) belongs to a family of tRNA synthases, of the class II enzymes. Class II tRNA synthases evolved early in evolution and are highly conserved. This is reflected by the fact that 498 of the 968-residue polypeptide human AARS shares 41% identity witht the E.coli protein. tRNA synthases are the enzymes that interpret the RNA code and attach specific aminoacids to the tRNAs that contain the cognate trinucleotide anticodons. They consist of a catalytic domain which interacts with the amino acid acceptor-T psi C helix of the tRNA, and a second domain which interacts with the rest of the tRNA structure.
Specifications
| DHX9 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Dhx9 | |
| AI326842; ATP-dependent RNA helicase A; Ddx9; D-E-A-D (aspartate-glutamate-alanine-aspartate) box polypeptide 9; DEAD (aspartate-glutamate-alanine-aspartate) box polypeptide 9; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 9; DEAH (Asp-Glu-Ala-His) box helicase 9; DEAH (Asp-Glu-Ala-His) box polypeptide 9; DEAH box protein 9; DEAH-box helicase 9; DExH-box helicase 9; Dhx9; FLJ17406; HEL-5; Leukophysin; LKP; mHEL-5; NDH II; NDH2; NDHII; Nuclear DNA helicase II; OTTHUMP00000033287; RHA; RNA helicase; RNA helicase A | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 13211, 1660, 304859 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB4140 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O70133, Q08211 | |
| Dhx9 | |
| A synthesized peptide derived from human RNA Helicase A (aa 1150-1200). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction