missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHX9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17721-1-AP-20UL
This item is not returnable.
View return policy
Description
The human alanyl-tRNA synthetase (AARS) belongs to a family of tRNA synthases, of the class II enzymes. Class II tRNA synthases evolved early in evolution and are highly conserved. This is reflected by the fact that 498 of the 968-residue polypeptide human AARS shares 41% identity witht the E.coli protein. tRNA synthases are the enzymes that interpret the RNA code and attach specific aminoacids to the tRNAs that contain the cognate trinucleotide anticodons. They consist of a catalytic domain which interacts with the amino acid acceptor-T psi C helix of the tRNA, and a second domain which interacts with the rest of the tRNA structure.Specifications
| DHX9 | |
| Polyclonal | |
| Unconjugated | |
| DHX9 | |
| ATP dependent RNA helicase A, DDX9, DEAH box protein 9, DHX9, LKP, NDH II, NDH2, NDHII, Nuclear DNA helicase II, RHA | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13211, 1660, 304859 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O70133, Q08211 | |
| DHX9 | |
| DHX9 Fusion Protein Ag12104 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction