Learn More
Invitrogen™ DHX9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116788
Description
Positive Samples: U-87MG, Mouse thymus, Mouse brain, Rat brain Immunogen sequence: VVPWSPPQSN WNPWTSSNID EGPLAFATPE QISMDLKNEL MYQLEQDHDL QAILQERELL PVKKFESEIL EAISQNSVVI IRGATGCGKT TQVPQFILDD FIQNDRAAEC NIVVTQPRRI SAVSVAERVA FERGEEPGKS CGYSVRFESI LPRPHASIMF CTVGVLLRKL EAGIRGISHV IVDEIHERDI NTDFLLVVLR DVVQAYPEVR IVLMSATIDT SMFCEYFFNC PIIEV.
The human alanyl-tRNA synthetase (AARS) belongs to a family of tRNA synthases, of the class II enzymes. Class II tRNA synthases evolved early in evolution and are highly conserved. This is reflected by the fact that 498 of the 968-residue polypeptide human AARS shares 41% identity witht the E.coli protein. tRNA synthases are the enzymes that interpret the RNA code and attach specific aminoacids to the tRNAs that contain the cognate trinucleotide anticodons. They consist of a catalytic domain which interacts with the amino acid acceptor-T psi C helix of the tRNA, and a second domain which interacts with the rest of the tRNA structure.
Specifications
| DHX9 | |
| Polyclonal | |
| Unconjugated | |
| Dhx9 | |
| AI326842; ATP-dependent RNA helicase A; Ddx9; D-E-A-D (aspartate-glutamate-alanine-aspartate) box polypeptide 9; DEAD (aspartate-glutamate-alanine-aspartate) box polypeptide 9; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 9; DEAH (Asp-Glu-Ala-His) box helicase 9; DEAH (Asp-Glu-Ala-His) box polypeptide 9; DEAH box protein 9; DEAH-box helicase 9; DExH-box helicase 9; Dhx9; FLJ17406; HEL-5; Leukophysin; LKP; mHEL-5; NDH II; NDH2; NDHII; Nuclear DNA helicase II; OTTHUMP00000033287; RHA; RNA helicase; RNA helicase A | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 13211, 1660, 304859 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.84 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| O70133, Q08211 | |
| Dhx9 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 329-563 of DHX9/RNA Helicase A (NP_0013482). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.