missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DHX8 Recombinant Rabbit Monoclonal Antibody (K01_1Q41)
Description
DHX8 Recombinant Monoclonal Antibody for Western Blot
This gene is a member of the DEAH box polypeptide family. The encoded protein contains the DEAH (Asp-Glu-Ala-His) motif which is characteristic of all DEAH box proteins, and is thought to function as an ATP-dependent RNA helicase that regulates the release of spliced mRNAs from spliceosomes prior to their export from the nucleus. This protein may be required for the replication of human immunodeficiency virus type 1 (HIV-1). Alternative splicing results in multiple transcript variants. [provided by RefSeq, Oct 2014].
Specifications
Specifications
| Antigen | DHX8 |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | K01_1Q41 |
| Concentration | 0.50 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 150mM NaCl, 40% glycerol, 0.05% BSA and 0.01% sodium azide; pH 7.4 |
| Gene | Dhx8 |
| Gene Accession No. | Q14562 |
| Gene Alias | ATP-dependent RNA helicase DHX8; DDX8; DEAD (aspartate-glutamate-alanine-aspartate) box polypeptide 8; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 8 (RNA helicase); DEAH (Asp-Glu-Ala-His) box polypeptide 8; DEAH box protein 8; DEAH-box helicase 8; DHX8; HRH1; LOW QUALITY PROTEIN: ATP-dependent RNA helicase DHX8; LRRGT00035; mDEAH6; mKIAA4096; PRP22; PRPF22; RNA helicase; RNA helicase HRH1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?