missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DHX8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101449
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total DHX8.
This gene is a member of the DEAH box polypeptide family. The encoded protein contains the DEAH (Asp-Glu-Ala-His) motif which is characteristic of all DEAH box proteins, and is thought to function as an ATP-dependent RNA helicase that regulates the release of spliced mRNAs from spliceosomes prior to their export from the nucleus. This protein may be required for the replication of human immunodeficiency virus type 1 (HIV-1). Alternative splicing results in multiple transcript variants. [provided by RefSeq, Oct 2014].
Specifications
| DHX8 | |
| Polyclonal | |
| Unconjugated | |
| Dhx8 | |
| ATP-dependent RNA helicase DHX8; DDX8; DEAD (aspartate-glutamate-alanine-aspartate) box polypeptide 8; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 8 (RNA helicase); DEAH (Asp-Glu-Ala-His) box polypeptide 8; DEAH box protein 8; DEAH-box helicase 8; DHX8; HRH1; LOW QUALITY PROTEIN: ATP-dependent RNA helicase DHX8; LRRGT00035; mDEAH6; mKIAA4096; PRP22; PRPF22; RNA helicase; RNA helicase HRH1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1659, 217207 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| A2A4P0, Q14562 | |
| Dhx8 | |
| A synthesized peptide derived from human DHX8(Accession Q14562), corresponding to amino acid residues K1032-M1082. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction