missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DHX57 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568742
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 87%; Dog: 93%; Guinea Pig: 100%; Horse: 79%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
Probable ATP-binding RNA helicase.
Specifications
| DHX57 | |
| Polyclonal | |
| Unconjugated | |
| DHX57 | |
| AW494914; DDX57; DEAH (Asp-Glu-Ala-Asp/His) box polypeptide 57; DEAH (Asp-Glu-Ala-Asp/His) box polypeptide 57 homolog; DEAH box protein 57; DEAH-box helicase 57; DEAH-box RNA/DNA helicase AAM73547; DExH-box helicase 57; DHX57; Putative ATP-dependent RNA helicase DHX57 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 90957 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q6P158 | |
| DHX57 | |
| synthetic peptide directed towards the C terminal region of human DHX57. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction