missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHX57 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92330-0.1ml
This item is not returnable.
View return policy
Description
DHX57 Polyclonal antibody specifically detects DHX57 in Human, Rat samples. It is validated for Western Blot
Specifications
| DHX57 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DDX57, DEAH (Asp-Glu-Ala-Asp/His) box polypeptide 57, DEAH box protein 57, DEAH-box RNA/DNA helicase AAM73547, EC 3.6.1, EC 3.6.4.13, FLJ32861, putative ATP-dependent RNA helicase DHX57 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 700-900 of human DHX57 (NP_945314.1). SATLNAELFSDYFNSCPVITIPGRTFPVDQFFLEDAIAVTRYVLQDGSPYMRSMKQISKEKLKARRNRTAFEEVEEDLRLSLHLQDQDSVKDAVPDQQLDFKQLLARYKGVSKSVIKTMSIMDFEKVNLELIEALLEWIVDGKHSYPPGAILVFLPGLAEIKMLYEQLQSNSLFNNRRSNRCVIHPLHSSLSSEEQQAVFV | |
| 0.1 mL | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 90957 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction