missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DHX40 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA546142
This item is not returnable.
View return policy
Description
Peptide sequence: KQLKDGISKD VLKKMQRRND DKSISDARAR FLERKQQRTQ DHSDTRKETG Sequence homology: Cow: 86%; Dog: 93%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 86%; Rat: 86%.
This gene encodes a member of the DExH/D box family of ATP-dependent RNA helicases that have an essential role in RNA metabolism. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 17.
Specifications
| DHX40 | |
| Polyclonal | |
| Unconjugated | |
| DHX40 | |
| 2410016C14Rik; ARG147; DDX40; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 40 (RNA helicase); DEAH (Asp-Glu-Ala-His) box polypeptide 40; DEAH box protein 40; DEAH-box helicase 40; Dhx40; PAD; probable ATP-dependent RNA helicase DHX40; Protein PAD | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 79665 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8IX18 | |
| DHX40 | |
| Synthetic peptide directed towards the C-terminal of human DHX40 (aa 730-779). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction