missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DHX38 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA565307

Product Code. 15928064

  • £476.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: IPVKDATSDLA IIARKGSQTV RKHREQKERK KAQHKHWELA GTKLGDIMGV KKEEEPDKAV TEDGKVDYRT EQKFADHMKR KSEASSEFAK KKSIL Highest antigen sequence identity to the following orthologs - mouse 97%, rat 97%.

This gene encodes a zinc finger transcription factor. The encoded protein likely plays a role in transcriptional repression of interleukin 2. Mutations in this gene have been associated with posterior polymorphous corneal dystrophy-3 and late-onset Fuchs endothelial corneal dystrophy. Alternatively spliced transcript variants encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DHX38
Polyclonal
Unconjugated
DHX38
5730550P09Rik; AI325984; ATP-dependent RNA helicase DHX38; AW540902; DDX38; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 38; DEAH (Asp-Glu-Ala-His) box polypeptide 38; DEAH box protein 38; DEAH-box helicase 38; DHX38; hPrp16; KIAA0224; mKIAA0224; pre-mRNA splicing factor 16; pre-mRNA splicing factor ATP-dependent RNA helicase PRP16; pre-mRNA-splicing factor ATP-dependent RNA helicase PRP16; PRP16; PRP16 homolog of S.cerevisiae; PRPF16
Rabbit
Antigen Affinity Chromatography
RUO
9785
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q92620
DHX38
Recombinant Human DHX38. Recombinant protein control fragment (Product #RP-104740).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.