missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DHX36 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100473
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total DHX36.
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp, are putative RNA helicases implicated in several cellular processes involving modifications of RNA secondary structure. DHX36 (DEAH box protein 36), also known as MLE-like protein 1 and RNA helicase associated with AU-rich element ARE (RHAU), belongs to RNA helicase of the DEAH family and may function in sex development and spermatogenesis. It is expressed in testis and is evolutionary conserved with true orthologs in almost all animal species. DHX36 plays a role in degradation and deadenylation of mRNAs containing in their 3'-UTR the consensus ARE sequence element. DHX36 is required for early embryogenesis.
Specifications
| DHX36 | |
| Polyclonal | |
| Unconjugated | |
| DHX36 | |
| 2810407E23Rik; AI452301; Asp-Glu-Ala-His; ATP-dependent DNA/RNA helicase DHX36; ATP-dependent RNA helicase DHX36; AU022184; Ddx36; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 36; DEAD/H box polypeptide 36; DEAH (Asp-Glu-Ala-His) box polypeptide 36; DEAH box protein 36; DEAH-box helicase 36; DEAH-box protein 36; DHX36; G4 resolvase-1; G4R1; G4-resolvase 1; G4-resolvase-1; KIAA1488; mKIAA1488; Mlel1; MLE-like protein 1; probable ATP-dependent RNA helicase DHX36; RHAU; RNA helicase associated with AU-rich element ARE; RNA helicase associated with AU-rich element protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 170506, 310461, 72162 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8VHK9, Q9H2U1 | |
| DHX36 | |
| A synthesized peptide derived from human DHX36(Accession Q9H2U1), corresponding to amino acid residues I942-K992. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction