missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHX36 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB200-353
This item is not returnable.
View return policy
Description
DHX36 Polyclonal specifically detects DHX36 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| DHX36 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| DDX36, DEAH (Asp-Glu-Ala-His) box polypeptide 36, DEAH box protein 36, EC 3.6.1, EC 3.6.4.13, G4 resolvase-1, KIAA1488G4R1, MLEL1RHAUDEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 36, MLE-like protein 1, probable ATP-dependent RNA helicase DHX36, RNA helicase associated with AU-rich element ARE | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 170506 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1:5000-1:15000, Immunohistochemistry, Immunoprecipitation 2 - 10 ug/mg lysate, Immunohistochemistry-Paraffin 1:1000 | |
| Q9H2U1 | |
| DHX36 | |
| The immunogen recognized by this antibody maps to a region between residue 950 and the C-terminus (residue 1008) of human DEAH (Asp-Glu-Ala-His) Box Polypeptide 36 using the numbering given in entry NP_065916.1 | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction