missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: SQEILSIAAM MQIQNIFVVP PNQKSHAIRV HRKFAVEEGD HLTMLNIYEA FIKHNKDSKW CQEHFLNYKG LVRAATVREQ LKKLLVKFQV PRKSSEGDP Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 89%.
May be involved in pre-mRNA splicing.
Specifications
Specifications
| Antigen | DHX35 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | DHX35 |
| Gene Accession No. | Q9H5Z1 |
| Gene Alias | 1200009D07Rik; C20orf15; DDX35; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 35; DEAH (Asp-Glu-Ala-His) box polypeptide 35; DEAH box protein 35; DEAH-box helicase 35; DEAH-box protein 35; Dhx35; KAIA0875; probable ATP-dependent RNA helicase DHX35 |
| Gene Symbols | DHX35 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
