missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHX32 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68306-1-IG-20UL
This item is not returnable.
View return policy
Description
The DHX32 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DHX32. This antibody recognizes human, mouse, rat antigen. The DHX32 antibody has been validated for the following applications: WB, ELISA, FC (Intra) analysis.Specifications
| DHX32 | |
| Monoclonal | |
| 1000 μg/mL | |
| DDX32; DHLP1; FLJ10694; FLJ10889 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 84 kDa | |
| Liquid |
| Western Blot, ELISA, Flow Cytometry (Intracellular Staining) | |
| 2A4G9 | |
| Unconjugated | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 55760 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction