Learn More
DHX30 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28814-1-AP-20UL
This item is not returnable.
View return policy
Description
The DHX30 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human DHX30. This antibody recognizes human antigen. The DHX30 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| DHX30 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 22907 | |
| IgG |
| Western Blot, ELISA | |
| 300 μg/mL | |
| Rabbit | |
| Primary | |
| Human |