missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DHX30 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541298
This item is not returnable.
View return policy
Description
Peptide sequence: AESGMAPGGP GEGDGSLVNA SRDLLKEFPQ PKNLLNSVIG RALGISHAKD Sequence homology: Cow: 93%; Dog: 93%; Human: 100%; Mouse: 93%; Pig: 93%; Rabbit: 93%.
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this DEAD box protein family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. DHX30 is a member of this family.DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this DEAD box protein family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a member of this family. The encoded protein has 97% sequence identity with the mouse HELG protein. Alternative splicing of this gene generates 3 transcript variants.
Specifications
| DHX30 | |
| Polyclonal | |
| Unconjugated | |
| DHX30 | |
| 2810477H02Rik; ATP-dependent RNA helicase DHX30; ATP-dependent RNA helicase DHX30; putative ATP-dependent RNA helicase DHX30; C130058C04Rik; Ddx30; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 30; DEAH (Asp-Glu-Ala-His) box helicase 30; DEAH (Asp-Glu-Ala-His) box polypeptide 30; DEAH box protein 30; DEAH-box helicase 30; DExH-box helicase 30; Dhx30; FLJ11214; Helg; KIAA0890; putative ATP-dependent RNA helicase DHX30; RETCOR; Ret-CoR; retina co-repressor; Unknown (protein for MGC:134480) | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 22907 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q7L2E3 | |
| DHX30 | |
| Synthetic peptide directed towards the N-terminal of human DHX30. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction