missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHX30 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £359.00
Specifications
| Antigen | DHX30 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18684442
|
Novus Biologicals
NBP2-92561-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18609861
|
Novus Biologicals
NBP2-92561-0.1ml |
0.1 mL |
£359.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DHX30 Polyclonal antibody specifically detects DHX30 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| DHX30 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Endocrinology | |
| PBS with 50% glycerol, pH7.3. | |
| 22907 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| ATP-dependent RNA helicase DHX30, DDX30, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 30, DEAH (Asp-Glu-Ala-His) box polypeptide 30, DEAH box protein 30, EC 3.6.1, EC 3.6.4.13, FLJ11214, KIAA0890RETCOR, putative ATP-dependent RNA helicase DHX30, retina co-repressor | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 906-1155 of human DHX30 (NP_055781.2). GWEEVLRWQDRSSRENYLEENLLYAPSLRFIHGLIKQFSENIYEAFLVGKPSDCTLASAQCNEYSEEEELVKGVLMAGLYPNLIQVRQGKVTRQGKFKPNSVTYRTKSGNILLHKSTINREATRLRSRWLTYFMAVKSNGSVFVRDSSQVHPLAVLLLTDGDVHIRDDGRRATISLSDSDLLRLEGDSRTVRLLKELRRALGRMVERSLRSELAALPPSVQEEHGQLLALLAELLRGPCGSFDVRKTADD | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title