missing translation for 'onlineSavingsMsg'
Learn More

DHX29 Antibody, Novus Biologicals™

Product Code. 18327929 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18327929 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18327929 Supplier Novus Biologicals Supplier No. H00054505B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

DHX29 Polyclonal antibody specifically detects DHX29 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHX29
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot
Formulation PBS (pH 7.4)
Gene Accession No. NP_061903
Gene Alias ATP-dependent RNA helicase DHX29, DDX29, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 29, DEAH (Asp-Glu-Ala-His) box polypeptide 29, DEAH box protein 29, EC 3.6.1, EC 3.6.4.13, FLJ21492, Nucleic acid helicase DDXx
Host Species Mouse
Immunogen DHX29 (NP_061903, 1 a.a. - 1369 a.a.) full-length human protein. MGGKNKKHKAPAAAVVRAAVSASRAKSAEAGIAGEAQSKKPVSRPATAAAAAAGSREPRVKQGPKIYSFNSTNDSSGPANLDKSILKVVINNKLEQRIIGVINEHKKQNNDKGMISGRLTAKKLQDLYMALQAFSFKTKDIEDAMTNTLLYGGDLHSALDWLCLNLSDDALPEGFSQEFEEQQPKSRPKFQSPQIQATISPPLQPKTKTYEEDPKSKPKKEEKNMEVNMKEWILRYAEQQNEEEKNENSKSLEEEEKFDPNERYLHLAAKLLDAKEQAATFKLEKNKQGQKEAQEKIRKFQREMETLEDHPVFNPAMKISHQQNERKKPPVATEGESALNFNLFEKSAAATEEEKDKKKEPHDVRNFDYTARSWTGKSPKQFLIDWVRKNLPKSPNPSFEKVPVGRYWKCRVRVIKSEDDVLVVCPTILTEDGMQAQHLGATLALYRLVKGQSVHQLLPPTYRDVWLEWSDAEKKREELNKMETNKPRDLFIAKLLNKLKQQQQQQQQHSENKRENSEDPEESWENLVSDEDFSALSLESANVEDLEPVRNLFRKLQSTPKYQKLLKERQQLPVFKHRDSIVETLKRHRVVVVAGETGSGKSTQVPHFLLEDLLLNEWEASKCNIVCTQPRRISAVSLANRVCDELGCENGPGGRNSLCGYQIRMESRACESTRLLYCTTGVLLRKLQEDGLLSNVSHVIVDEVHERSVQSDFLLIILKEILQKRSDLHLILMSATVDSEKFSTYFTHCPILRISGRSYPVEVFHLEDIIEETGFVLEKDSEYCQKFLEEEEEVTINVTSKAGGIKKYQEYIPVQTGAHADLNPFYQKYSSRTQHAILYMNPHKINLDLILELLAYLDKSPQFRNIEGAVLIFLPGLAHIQQLYDLLSNDRRFYSERYKVIALHSILSTQDQAAAFTLPPPGVRKIVLATNIAETGITIPDVVFVIDTGRTKENKYHESSQMSSLVETFVSKASALQRQGRAGRVRDGFCFRMYTRERFEGFMDYSVPEILRVPLEELCLHIMKCNLGSPEDFLSKALDPPQLQVISNAMNLLRKIGACELNEPKLTPLGQHLAALPVNVKIGKMLIFGAIFGCLDPVATLAAVMTEKSPFTTPIGRKDEADLAKSALAMADSDHLTIYNAYLGWKKARQEGGYRSEITYCRRNFLNRTSLLTLEDVKQELIKLVKAAGFSSSTTSTSWEGNRASQTLSFQEIALLKAVLVAGLYDNVGKIIYTKSVDVTEKLACIVETAQGKAQVHPSSVNRDLQTHGWLLYQEKIRYARVYLRETTLITPFPVLLFGGDIEVQHRERLLSIDGWIYFQAPVKIAVIFKQLRVLIDSVLRKKLENPKMSLENDKILQIITELIKTENN
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Chromatin Research
Primary or Secondary Primary
Gene ID (Entrez) 54505
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.