missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHX29 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92373-0.02ml
This item is not returnable.
View return policy
Description
DHX29 Polyclonal antibody specifically detects DHX29 in Human samples. It is validated for Western Blot
Specifications
| DHX29 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| ATP-dependent RNA helicase DHX29, DDX29, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 29, DEAH (Asp-Glu-Ala-His) box polypeptide 29, DEAH box protein 29, EC 3.6.1, EC 3.6.4.13, FLJ21492, Nucleic acid helicase DDXx | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1020-1369 of human DHX29 (NP_061903.2). LHIMKCNLGSPEDFLSKALDPPQLQVISNAMNLLRKIGACELNEPKLTPLGQHLAALPVNVKIGKMLIFGAIFGCLDPVATLAAVMTEKSPFTTPIGRKDEADLAKSALAMADSDHLTIYNAYLGWKKARQEGGYRSEITYCRRNFLNRTSLLTLEDVKQELIKLVKAAGFSSSTTSTSWEGNRASQTLSFQEIALLKAVLVAGLYDNVGKIIYTKSVDVTEKLACIVETAQGKAQVHPSSVNRDLQTHGWLLYQEKIRYARVYLRETTLITPFPVLLFGGDIEVQHRERLLSIDGWIYFQAPVKIAVIFKQLRVLIDSVLRKKLENPKMSLENDKILQIITELIKTENN | |
| 0.02 mL | |
| Chromatin Research | |
| 54505 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction