missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DHX16 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541404
This item is not returnable.
View return policy
Description
Peptide sequence: KYQLVLEEEE TIEFVRATQL QGDEEPSAPP TSTQAQQKES IQAVRRSLPV Sequence homology: Cow: 86%; Goat: 86%; Horse: 86%; Human: 100%; Mouse: 79%; Pig: 85%; Rabbit: 77%; Rat: 79%.
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which is a functional homolog of fission yeast Prp8 protein involved in cell cycle progression. This gene is mapped to the MHC region on chromosome 6p21.3, a region where many malignant, genetic and autoimmune disease genes are linked. Three transcript variants encoding different isoforms have been found for this gene.
Specifications
| DHX16 | |
| Polyclonal | |
| Unconjugated | |
| Dhx16 | |
| 2410006N22Rik; ATP-dependent RNA helicase #3; DBP2; Ddx16; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 16; DEAD/H box 16; DEAH (Asp-Glu-Ala-His) box polypeptide 16; DEAH-box helicase 16; DEAH-box protein 16; Dhx16; KIAA0577; mKIAA0577; Pre-mRNA-splicing factor ATP-dependent RNA helicase DHX16; pre-mRNA-splicing factor ATP-dependent RNA helicase DHX16; putative pre-mRNA-splicing factor ATP-dependent RNA helicase DHX16; PRO2014; Prp2; PRP8; PRPF2; putative pre-mRNA-splicing factor ATP-dependent RNA helicase DHX16; RNA helicase | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 8449 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O60231 | |
| Dhx16 | |
| Synthetic peptide directed towards the N-terminal of human DHX16 (aa 352-401). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction