missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHX16 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-87270
This item is not returnable.
View return policy
Description
DHX16 Polyclonal specifically detects DHX16 in Human samples. It is validated for Western Blot.
Specifications
| DHX16 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| ATP-dependent RNA helicase #3, DBP2PRO2014, DDX16DEAD/H box 16, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 16, DEAH (Asp-Glu-Ala-His) box polypeptide 16, DEAH-box protein 16, EC 3.6.1, EC 3.6.4.13, KIAA0577, Prp2, PRP8, PRPF2, putative pre-mRNA-splicing factor ATP-dependent RNA helicase DHX16, RNA helicase | |
| The immunogen is a synthetic peptide directed towards the N-terminal region of DHX16 | |
| 100 μL | |
| Cell Cycle and Replication | |
| 8449 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction