missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DHX15 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561413
This item is not returnable.
View return policy
Description
Immunogen sequence: RASTNAMLIS AGLPPLKASH SAHSTHSAHS THSTHSAHST HAGHAGHTSL PQCINPFTNL PHTPRYYDIL KKRLQLPVWE YKD Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 99%.
DHX15 (DEAH-Box Helicase 15) is a Protein Coding gene. Among its related pathways are Gene Expression and mRNA Splicing - Major Pathway. Gene Ontology (GO) annotations related to this gene include nucleic acid binding and helicase activity. An important paralog of this gene is DHX8.
Specifications
| DHX15 | |
| Polyclonal | |
| Unconjugated | |
| DHX15 | |
| ATP-dependent RNA helicase #46; DBP1; DDX15; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 15; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 15 (RNA helicase A); DEAD/H box-15; DEAH (Asp-Glu-Ala-His) box helicase 15; DEAH (Asp-Glu-Ala-His) box polypeptide 15; DEAH box protein 15; DEAH9; DEAH-box helicase 15; dhx15; HRH2; im:2639158; mDEAH9; Pre-mRNA-splicing factor ATP-dependent RNA helicase DHX15; PRP43; PRPF43; PrPp43p; putative pre-mRNA-splicing factor ATP-dependent RNA helicase DHX15; putative pre-mRNA-splicing factor ATP-dependent RNA helicase DHX15-like protein; RNA helicase 2; wu:fb38f09; wu:fk62f05 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 13204, 1665, 289693 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O35286, O43143 | |
| DHX15 | |
| Recombinant protein corresponding to Human DHX15. Recombinant protein control fragment (Product #RP-100115). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction