missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHTKD1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27493-1-AP-150UL
This item is not returnable.
View return policy
Description
The 2-oxoglutarate dehydrogenase complex catalyzes the overall conversion of 2-oxoglutarate to succinyl-CoA and CO(2). It contains multiple copies of three enzymatic components: 2-oxoglutarate dehydrogenase (E1), dihydrolipoamide succinyltransferase (E2) and lipoamide dehydrogenase (E3).Specifications
| DHTKD1 | |
| Polyclonal | |
| Unconjugated | |
| DHTKD1 | |
| DHTKD1, KIAA1630 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55526 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q96HY7 | |
| DHTKD1 | |
| DHTKD1 Fusion Protein Ag26515 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction