missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DHTKD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117910
This item is not returnable.
View return policy
Description
DHTKD1 Polyclonal Antibody for Western Blot, IP
The 2-oxoglutarate dehydrogenase complex catalyzes the overall conversion of 2-oxoglutarate to succinyl-CoA and CO(2). It contains multiple copies of three enzymatic components: 2-oxoglutarate dehydrogenase (E1), dihydrolipoamide succinyltransferase (E2) and lipoamide dehydrogenase (E3).
Specifications
| DHTKD1 | |
| Polyclonal | |
| PBS with no preservative | |
| Q96HY7 | |
| DHTKD1 | |
| A synthetic peptide corresponding to the N-terminus of the Human DHTKD1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| DHTKD1 | |
| AMOXAD; C330018I04Rik; CMT2Q; dehydrogenase E1 and transketolase domain containing 1; dehydrogenase E1 and transketolase domain-containing protein 1; Dhtkd1; DKFZp762M115; KIAA1630; MGC3090; Probable 2-oxoglutarate dehydrogenase E1 component DHKTD1, mitochondrial | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 55526 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction