missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DHTKD1 Polyclonal specifically detects DHTKD1 in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | DHTKD1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 750 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | dehydrogenase E1 and transketolase domain containing 1, Dehydrogenase E1 and transketolase domain-containing protein 1, DKFZp762M115, EC 1.2.4.2, KIAA1630DKFZP762M115, MGC3090, probable 2-oxoglutarate dehydrogenase E1 component DHKTD1, mitochondrial |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human DHTKD1. |
| Purification Method | Antigen Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?