missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHRSX Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA551803
This item is not returnable.
View return policy
Description
Immunogen sequence: PRPDRVAIVT GGTDGIGYST AKHLARLGMH VIIAGNNDSK AKQVVSKIKE ETLNDK Highest antigen sequence identity to the following orthologs: Mouse - 57%, Rat - 54%.
DHRSX (dehydrogenase/reductase SDR family member on chromosome X) is a 330 amino acid protein belonging to the short-chain dehydrogenases/reductases (SDR) family. Widely expressed, DHRSX is an oxidoreductase that contains a coenzyme binding site and a substrate binding site, indicating a possible role in cellular metabolism. The gene that encodes DHRSX is located in the pseudoautosomal region 1 (PAR1) of X and Y chromosomes. The X and Y chromosomes are the human sex chromosomes. Chromosome X consists of about 153 million base pairs and nearly 1,000 genes.
Specifications
| DHRSX | |
| Polyclonal | |
| Unconjugated | |
| DHRSX | |
| CXorf11, DHRS5X, SDR46C1, SDR7C6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 207063 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N5I4 | |
| DHRSX | |
| Recombit protein corresponding to Human DHRSX | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction