missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DHRS9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103782
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total DHRS9.
DHRS9 is a 3-alpha-hydroxysteroid dehydrogenase that converts 3-alpha-tetrahydroprogesterone (allopregnanolone) to dihydroxyprogesterone and 3-alpha-androstanediol to dihydroxyprogesterone.
Specifications
| DHRS9 | |
| Polyclonal | |
| Unconjugated | |
| Dhrs9 | |
| 3-alpha hydroxysteroid dehydrogenase; 3-alpha-HSD; 3alpha-HSD; C730025I08Rik; dehydrogenase/reductase (SDR family) member 9; dehydrogenase/reductase 9; dehydrogenase/reductase SDR family member 9; DHRS9; epithelial retinol dehydrogenase; eRolDH; NADP-dependent retinol dehydrogenase/reductase; RDH15; RDH-E2; RDHL; RDHTBE; RDH-TBE; Retinol dehydrogenase; retinol dehydrogenase 15; retinol dehydrogenase homolog; RETSDR8; SDR9C4; Short chain dehydrogenase/reductase family 9C member 4; short chain dehydrogenase/reductase family 9C, member 4; short-chain dehydrogenase/reductase retSDR8; Tracheobronchial epithelial cell-specific retinol dehydrogenase; UNQ835/PRO1773 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10170, 170635, 241452 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q58NB6, Q8VD48, Q9BPW9 | |
| Dhrs9 | |
| A synthesized peptide derived from human DHRS9(Accession Q9BPW9), corresponding to amino acid residues S164-A214. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction