missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DHRS9 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17100024 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17100024 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17100024 Supplier Invitrogen™ Supplier No. PA5112158

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This product can be used in direct ELISA.

DHRS9 is a 3-alpha-hydroxysteroid dehydrogenase that converts 3-alpha-tetrahydroprogesterone (allopregnanolone) to dihydroxyprogesterone and 3-alpha-androstanediol to dihydroxyprogesterone.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHRS9
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Dhrs9
Gene Accession No. Q9BPW9
Gene Alias 3-alpha hydroxysteroid dehydrogenase; 3-alpha-HSD; 3alpha-HSD; C730025I08Rik; dehydrogenase/reductase (SDR family) member 9; dehydrogenase/reductase 9; dehydrogenase/reductase SDR family member 9; DHRS9; epithelial retinol dehydrogenase; eRolDH; NADP-dependent retinol dehydrogenase/reductase; RDH15; RDH-E2; RDHL; RDHTBE; RDH-TBE; Retinol dehydrogenase; retinol dehydrogenase 15; retinol dehydrogenase homolog; RETSDR8; SDR9C4; Short chain dehydrogenase/reductase family 9C member 4; short chain dehydrogenase/reductase family 9C, member 4; short-chain dehydrogenase/reductase retSDR8; Tracheobronchial epithelial cell-specific retinol dehydrogenase; UNQ835/PRO1773
Gene Symbols Dhrs9
Host Species Rabbit
Immunogen Recombinant Human DHRS9/3-alpha-HSD Protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10170
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.