missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHRS9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£178.00 - £255.00
Specifications
| Antigen | DHRS9 |
|---|---|
| Dilution | Western Blot 1:500-1:1000, ELISA 1:25000-1:50000, Immunoprecipitation 0.2-1 μL/mg of lysate |
| Applications | Western Blot, ELISA, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18656952
|
Novus Biologicals
NBP2-97017-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18689841
|
Novus Biologicals
NBP2-97017-50ul |
50 μg |
£178.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DHRS9 Polyclonal antibody specifically detects DHRS9 in Human samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
| DHRS9 | |
| Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| metabolism | |
| 0.2 um filtered solution in PBS | |
| 10170 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:1000, ELISA 1:25000-1:50000, Immunoprecipitation 0.2-1 μL/mg of lysate | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| dehydrogenase/reductase (SDR family) member 9,3alpha-HSD, EC 1.1, EC 1.1.1, EC 1.1.1.105, member 4,3-alpha-HSD, NADP-dependent retinol dehydrogenase/reductase, RDH15, RDH-E2, RDHL3ALPHA-HSD, RDHTBE | |
| Produced in rabbits immunized with purified, recombinant Human DHRS9 (Accession#: NP_005762.2; Arg18-Val319) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title