missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHRS9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38244-100ul
This item is not returnable.
View return policy
Description
DHRS9 Polyclonal antibody specifically detects DHRS9 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| DHRS9 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:10 - 1:100 | |
| dehydrogenase/reductase (SDR family) member 9,3alpha-HSD, EC 1.1, EC 1.1.1, EC 1.1.1.105, member 4,3-alpha-HSD, NADP-dependent retinol dehydrogenase/reductase, RDH15, RDH-E2, RDHL3ALPHA-HSD, RDHTBE | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 18-319 of human DHRS9 (NP_001135743.1).,, Sequence:, RKGKLKIEDITDKYIFITGCDSGFGNLAARTFDKKGFHVIAACLTESGSTALKAETSERLRTVLLDVTDPENVKRTAQWVKNQVGEKGLWGLINNAGVPGVLAPTDWLTLEDYREPIEVNLFGLISVTLNMLPLVKKAQGRVINVSSVGGRLAIVGGGYTPSKYAVEGFNDSLRRDMKAFGVHVSCIEPGLFKTNLADPVKVIEKKLAIWEQLSPDIKQQYGEGYIEKSLDKLKGNKSYVNMDLSPVVECMDHALTSLFPKTHYAAGKDAKIFWIPLSHMPAALQDFLLLKQKAELANPKAV | |
| 100 μL | |
| metabolism | |
| 10170 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction