missing translation for 'onlineSavingsMsg'
Learn More

DHRS7B Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Thermo Scientific PA552916

Additional Details : Weight : 0.02000kg

Product Code. 15790861

  • £427.00 / 100µL
Estimated Shipment: 21-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Immunogen sequence: ATQAFFDCLR AEMEQYEIEV TVISPGYIHT NLSVNAITAD GSRYGVMDTT TAQGRSPVEV AQDVLAAVGK KKKDVILADL LPSLAVYLRT LAPGLFFSLM ASRARKERKS KNS Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 81%.

DHRS7B Polyclonal antibody specifically detects DHRS7B in Human samples. It is validated for Immunohistochemistry, Western Blot.
Specifications

Specifications

DHRS7B
Polyclonal
Unconjugated
DHRS7B
BC003479; C79874; C80074; CGI-93; dehydrogenase/reductase (SDR family) member 7B; dehydrogenase/reductase 7B; dehydrogenase/reductase SDR family member 7B; Dhrs7b; PexRAP; RGD1311243; SDR family dehydrogenase/reductase member 7B; SDR32C1; short chain dehydrogenase/reductase family 32C, member 1; short-chain dehydrogenase/reductase family 32C member 1; UNQ212/PRO238
Rabbit
Antigen affinity chromatography
RUO
25979
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry, Western Blot
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q6IAN0
DHRS7B
Recombinant protein corresponding to Human DHRS7B
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Research Use Only