missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DHRS7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569451
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 92%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 92%.
Short-chain dehydrogenases/reductases (SDRs), such as DHRS7, catalyze the oxidation/reduction of a wide range of substrates, including retinoids and steroids (Haeseleer and Palczewski, 2000 [PubMed 10800688]).[supplied by OMIM, Jun 2009].
Specifications
| DHRS7 | |
| Polyclonal | |
| Unconjugated | |
| DHRS7 | |
| 2310016E22Rik; 5730564L20Rik; AW061210; CGI-86; dehydrogenase/reductase (SDR family) member 7; dehydrogenase/reductase 7; dehydrogenase/reductase SDR family member 7; Dhrs7; DHRS7A; Retdsr4; retinal short-chain dehydrogenase/reductase 4; Retsdr4; RGD1565002; SDR34C1; short chain dehydrogenase/reductase family 34C member 1; short chain dehydrogenase/reductase family 34C, member 1; similar to Dehydrogenase/reductase SDR family member 7 precursor (Retinal short-chain dehydrogenase/reductase 4); similar to Dehydrogenase/reductase SDR family member 7 precursor (Retinal short-chain dehydrogenase/reductase 4) (retSDR4); uncharacterized protein LOC299135; UNQ285/PRO3448 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 66375 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9CXR1 | |
| DHRS7 | |
| synthetic peptide corresponding to a region of mouse DHRS7 (aa 51-100). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction