Learn More
Invitrogen™ DHRS7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556649
Description
Immunogen sequence: DLTLLWAEWQ GRRPEWELTD MVVWVTGASS GIGEELAYQL SKLGVSLVLS ARRVHELERV KRRCLENGNL KEKDILVLPL DLTDTGSHEA ATKAVLQEFG RIDILVNNGG MSQRSLCMDT SLDVYRKLIE LNYLGTVSLT KCVLPHMI Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 85%.
Short-chain dehydrogenases/reductases (SDRs), such as DHRS7, catalyze the oxidation/reduction of a wide range of substrates, including retinoids and steroids (Haeseleer and Palczewski, 2000 [PubMed 10800688]).[supplied by OMIM, Jun 2009].
Specifications
| DHRS7 | |
| Polyclonal | |
| Unconjugated | |
| DHRS7 | |
| 2310016E22Rik; 5730564L20Rik; AW061210; CGI-86; dehydrogenase/reductase (SDR family) member 7; dehydrogenase/reductase 7; dehydrogenase/reductase SDR family member 7; Dhrs7; DHRS7A; Retdsr4; retinal short-chain dehydrogenase/reductase 4; Retsdr4; RGD1565002; SDR34C1; short chain dehydrogenase/reductase family 34C member 1; short chain dehydrogenase/reductase family 34C, member 1; similar to Dehydrogenase/reductase SDR family member 7 precursor (Retinal short-chain dehydrogenase/reductase 4); similar to Dehydrogenase/reductase SDR family member 7 precursor (Retinal short-chain dehydrogenase/reductase 4) (retSDR4); uncharacterized protein LOC299135; UNQ285/PRO3448 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51635 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y394 | |
| DHRS7 | |
| Recombinant protein corresponding to Human DHRS7. Recombinant protein control fragment (Product #RP-88890). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.