missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DHRS7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA532720
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Gibbon (100%) Monkey (95%) Pig (89%).
Short-chain dehydrogenases/reductases (SDRs), such as DHRS7, catalyze the oxidation/reduction of a wide range of substrates, including retinoids and steroids (Haeseleer and Palczewski, 2000 [PubMed 10800688]).[supplied by OMIM, Jun 2009].
Specifications
| DHRS7 | |
| Polyclonal | |
| Unconjugated | |
| DHRS7 | |
| 2310016E22Rik; 5730564L20Rik; AW061210; CGI-86; dehydrogenase/reductase (SDR family) member 7; dehydrogenase/reductase 7; dehydrogenase/reductase SDR family member 7; Dhrs7; DHRS7A; Retdsr4; retinal short-chain dehydrogenase/reductase 4; Retsdr4; RGD1565002; SDR34C1; short chain dehydrogenase/reductase family 34C member 1; short chain dehydrogenase/reductase family 34C, member 1; similar to Dehydrogenase/reductase SDR family member 7 precursor (Retinal short-chain dehydrogenase/reductase 4); similar to Dehydrogenase/reductase SDR family member 7 precursor (Retinal short-chain dehydrogenase/reductase 4) (retSDR4); uncharacterized protein LOC299135; UNQ285/PRO3448 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51635 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q9Y394 | |
| DHRS7 | |
| Synthetic 19 amino acid peptide from C-terminus of human DHRS7. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction