missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DHRS4L2 Monoclonal Antibody (OTI8D1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527281
This item is not returnable.
View return policy
Description
DHRS4L2 Monoclonal Antibody for Western Blot, IHC (P)
DHRS4L2 encodes a member of the short chain dehydrogenase reductase family. The encoded protein may be an NADPH dependent retinol oxidoreductase. Alternate splicing results in multiple transcript variants.
Specifications
| DHRS4L2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6PKH6 | |
| DHRS4L2 | |
| Full length human recombinant protein of DHRS4L2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI8D1 | |
| Unconjugated | |
| DHRS4L2 | |
| dehydrogenase/reductase (SDR family) member 4 like 2; dehydrogenase/reductase (SDR family) member 4 like 2A3; dehydrogenase/reductase 4 like 2; dehydrogenase/reductase SDR family member 4-like 2; DHRS4L2; NADP(H)-dependent retinol dehydrogenase/reductase short isoform-like protein; NADPH-dependent retinol dehydrogenase/reductase-like protein 2; SDR25C3; short chain dehydrogenase/reductase family 25C member 3; short chain dehydrogenase/reductase family 25C, member 3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 317749 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction