missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DHRS4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115119
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total DHRS4.
Reduces all-trans-retinal and 9-cis retinal. Can also catalyze the oxidation of all-trans-retinol with NADP as co-factor, but with much lower efficiency. Reduces alkyl phenyl ketones and alpha-dicarbonyl compounds with aromatic rings, such as pyrimidine-4-aldehyde, 3-benzoylpyridine, 4-benzoylpyridine, menadione and 4-hexanoylpyridine. Has no activity towards aliphatic aldehydes and ketones.
Specifica
| DHRS4 | |
| Polyclonal | |
| Unconjugated | |
| DHRS4 | |
| AI043103; AI790593; carbonyl reductase; CR; D14Ucla2; DCR-AKL; dehydrogenase/reductase (SDR family) member 4; dehydrogenase/reductase (SDR family) member 4 like 2A; dehydrogenase/reductase 4; dehydrogenase/reductase SDR family member 4; DHRS4; mouNRDR; NADH-dependent retinal reductase; NADPH-dependent carbonyl reductase/NADP-retinol dehydrogenase; NADPH-dependent retinol dehydrogenase/reductase; NADP-retinol dehydrogenase; NDRD; NDRD protein; NRDR; Peroxisomal short-chain alcohol dehydrogenase; PH PSCD; PHCR; PSCD; rabNRDR; RRD; SCAD-SRL; SDR25C1; SDR25C2; SDR-SRL; Short chain dehydrogenase/reductase family 25C member 2; short chain dehydrogenase/reductase family 25C, member 1; short chain dehydrogenase/reductase family 25C, member 2; short-chain dehydrogenase/reductase family member 4; UNQ851/PRO1800 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10901, 266686, 28200 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q8VID1, Q99LB2, Q9BTZ2 | |
| DHRS4 | |
| A synthesized peptide derived from human DHRS4(Accession Q9BTZ2), corresponding to amino acid residues C209-D259. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto