missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DHRS4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554943
This item is not returnable.
View return policy
Description
Immunogen sequence: SIAAFSPSPG FSPYNVSKTA LLGLTKTLAI ELAPRNIRVN CLAPGLIKTS FSRMLWMDKE KEESMKETLR IRRLGEPEDC AGIVSFLCSE DASYITGETV VVGGGT Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 75%.
Reduces all-trans-retinal and 9-cis retinal. Can also catalyze the oxidation of all-trans-retinol with NADP as co-factor, but with much lower efficiency. Reduces alkyl phenyl ketones and alpha-dicarbonyl compounds with aromatic rings, such as pyrimidine-4-aldehyde, 3-benzoylpyridine, 4-benzoylpyridine, menadione and 4-hexanoylpyridine. Has no activity towards aliphatic aldehydes and ketones.
Specifications
| DHRS4 | |
| Polyclonal | |
| Unconjugated | |
| DHRS4 | |
| AI043103; AI790593; carbonyl reductase; CR; D14Ucla2; DCR-AKL; dehydrogenase/reductase (SDR family) member 4; dehydrogenase/reductase (SDR family) member 4 like 2A; dehydrogenase/reductase 4; dehydrogenase/reductase SDR family member 4; DHRS4; mouNRDR; NADH-dependent retinal reductase; NADPH-dependent carbonyl reductase/NADP-retinol dehydrogenase; NADPH-dependent retinol dehydrogenase/reductase; NADP-retinol dehydrogenase; NDRD; NDRD protein; NRDR; Peroxisomal short-chain alcohol dehydrogenase; PH PSCD; PHCR; PSCD; rabNRDR; RRD; SCAD-SRL; SDR25C1; SDR25C2; SDR-SRL; Short chain dehydrogenase/reductase family 25C member 2; short chain dehydrogenase/reductase family 25C, member 1; short chain dehydrogenase/reductase family 25C, member 2; short-chain dehydrogenase/reductase family member 4; UNQ851/PRO1800 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10901 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BTZ2 | |
| DHRS4 | |
| Recombinant protein corresponding to Human DHRS4. Recombinant protein control fragment (Product #RP-89060). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction