missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DHRS3 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA552750

Product Code. 15740701

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: TEKCLKETTE EIRQMGTECH YFICDVGNRE EVYQTAKAVR EKVGDITILV NNAAVVHGKS LMDSDDDALL KSQHINTLGQ FWTTKAFLPR MLELQNGHIV CLNSVLALSA IPG Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

DHRS3 catalyze the oxidation/reduction of a wide range of substrates, including retinoids and steroids.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DHRS3
Polyclonal
Unconjugated
Dhrs3
DD83.1; dehydrogenase/reductase (SDR family) member 3; dehydrogenase/reductase 3; dehydrogenase/reductase member 3; Dhrs3; RDH17; retinal dehydrogenase/reductase family member 3; retinal short-chain dehydrogenase/reductase 1; Retinol dehydrogenase 17; retSDR1; Rsdr1; SDR1; SDR16C1; Short chain dehydrogenase/reductase family 16C member 1; short chain dehydrogenase/reductase family 16C, member 1; short-chain dehydrogenase/reductase 1; short-chain dehydrogenase/reductase 3; UNQ2424/PRO4983
Rabbit
Antigen affinity chromatography
RUO
20148, 313689, 9249
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O75911, O88876
Dhrs3
Recombinant protein corresponding to Human DHRS3. Recombinant protein control fragment (Product #RP-90377).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.