missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DHRS2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565828
This item is not returnable.
View return policy
Description
Immunogen sequence: EDREQLVAKAL EHCGGVDFLV CSAGVNPLVG STLGTSEQIW DKILSVNVKS PALLLSQL Highest antigen sequence identity to the following orthologs - mouse 80%, rat 80%.
DHRS2 displays NADPH-dependent dicarbonyl reductase activity in vitro with 3,4-Hexanedione, 2,3-Heptanedione and 1-Phenyl-1,2-propanedione as substrates. DHRS2 do not reductase activity is displayed in vitro with steroids, retinoids and sugars as substrates. This protein may inhibit cell replication.
Specifications
| DHRS2 | |
| Polyclonal | |
| Unconjugated | |
| DHRS2 | |
| 5430405K24Rik; dehydrogenase/reductase (SDR family) member 2; dehydrogenase/reductase 2; dehydrogenase/reductase member 2; dehydrogenase/reductase SDR family member 2, mitochondrial; Dhrs2; Dicarbonyl reductase HEP27; Hep27; Protein D; SDR family member; SDR25C1; Short chain dehydrogenase/reductase family 25C member 1; short chain dehydrogenase/reductase family 25C, member 1; short-chain alcohol dehydrogenase family member | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10202 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q13268 | |
| DHRS2 | |
| Recombinant Human DHRS2. Recombinant protein control fragment (Product #RP-104146). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction