missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DHPS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553170
This item is not returnable.
View return policy
Description
Immunogen sequence: GKELRENGIN RIGNLLVPNE NYCKFEDWLM PILDQMVMEQ NTEGVKWTPS KMIARLGKEI NNPESVYYWA QKNHIPVFSP ALTDGSLGDM IFFHSYKNPG LVLDIVE Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 95%.
This gene encodes a protein that is required for the formation of hypusine, a unique amino acid formed by the posttranslational modification of only one protein, eukaryotic translation initiation factor 5A. The encoded protein catalyzes the first step in hypusine formation by transferring the butylamine moiety of spermidine to a specific lysine residue of the eukaryotic translation initiation factor 5A precursor, forming an intermediate deoxyhypusine residue. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications
| DHPS | |
| Polyclonal | |
| Unconjugated | |
| Dhps | |
| Deoxyhypusine synthase; DHPS; DHS; DS; MIG13; migration-inducing gene 13 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1725 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P49366 | |
| Dhps | |
| Recombinant protein corresponding to Human DHPS. Recombinant protein control fragment (Product #RP-89630). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction