missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHPS Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32256UV
This item is not returnable.
View return policy
Description
DHPS Polyclonal antibody specifically detects DHPS in Human, Mouse samples. It is validated for Western Blot
Specifications
| DHPS | |
| Polyclonal | |
| Western Blot | |
| deoxyhypusine synthase, DHS, DS, EC 2.5.1, EC 2.5.1.46, hypusine, MIG13, migration-inducing gene 13 | |
| The immunogen this antibody was made to, maps to a region between residue 319 to 369 of human Deoxyhypusine Synthase using the numbering given in entry NP_001921.1 (GeneID 1725). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 1725 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction