missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DHFRL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562412
This item is not returnable.
View return policy
Description
Immunogen sequence: PLRNEFRYFQ RMTTTSSVEG KQNLVIMGRK Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 93%.
Key enzyme in folate metabolism. Contributes to the de novo mitochondrial thymidylate biosynthesis pathway. Required to prevent uracil accumulation in mtDNA. Binds its own mRNA and that of DHFR.
Specifications
| DHFRL1 | |
| Polyclonal | |
| Unconjugated | |
| DHFR2 | |
| DHFR2; DHFRL1; DHFRP4; dihydrofolate reductase 2; Dihydrofolate reductase 2, mitochondrial; dihydrofolate reductase like 1; dihydrofolate reductase pseudogene 4; dihydrofolate reductase, mitochondrial; dihydrofolate reductase-like 1; dihydrofolate reductase-like protein 1; LOW QUALITY PROTEIN: dihydrofolate reductase 2, mitochondrial | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 200895 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86XF0 | |
| DHFR2 | |
| Recombinant protein corresponding to Human DHFRL1. Recombinant protein control fragment (Product #RP-99284). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction