missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHFRL1 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00200895-B01P
This item is not returnable.
View return policy
Description
DHFRL1 Polyclonal antibody specifically detects DHFRL1 in Human samples. It is validated for Western Blot
Specifications
| DHFRL1 | |
| Polyclonal | |
| PBS (pH 7.4) | |
| DHFRP4, dihydrofolate reductase pseudogene 4, dihydrofolate reductase-like 1, dihydrofolate reductase-like protein 1, EC 1.5.1.-, EC 1.5.1.3, FLJ16119 | |
| DHFRL1 (NP_789785.1, 1 a.a. - 187 a.a.) full-length human protein. MFLLLNCIVAVSQNMGIGKNGDLPRPPLRNEFRYFQRMTTTSSVEGKQNLVIMGRKTWFSIPEKNRPLKDRINLVLSRELKEPPQGAHFLARSLDDALKLTERPELANKVDMIWIVGGSSVYKEAMNHLGHLKLFVTRIMQDFESDTFFSEIDLEKYKLLPEYPGVLSDVQEGKHIKYKFEVCEKDD | |
| 0.05 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| NP_789785.1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 200895 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction