missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHFR Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
£119.00 - £295.00
Specifications
| Antigen | DHFR |
|---|---|
| Clone | 4C11E9 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Flow Cytometry (Intracellular Staining), ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30237392
|
Proteintech
68274-1-IG-20UL |
20 μL |
£135.00 £119.00 / 20µL Save £16.00 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30236048
|
Proteintech
68274-1-IG-150UL |
150 μL |
£349.00 £295.00 / 150µL Save £54.00 15% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descripción
The DHFR antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DHFR. This antibody recognizes human, mouse antigen. The DHFR antibody has been validated for the following applications: WB, FC (Intra), ELISA analysis.Especificaciones
| DHFR | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 1719 | |
| IgG1 | |
| Protein G purification | |
| Monoclonal Antibody |
| 4C11E9 | |
| Western Blot, Flow Cytometry (Intracellular Staining), ELISA | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto