missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHDH Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-16158
This item is not returnable.
View return policy
Description
DHDH Polyclonal specifically detects DHDH in Human samples. It is validated for Western Blot.
Specifications
| DHDH | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| 3-deoxyglucosone reductase, dihydrodiol dehydrogenase (dimeric), Dimeric dihydrodiol dehydrogenase, D-xylose 1-dehydrogenase, D-xylose-NADP dehydrogenase, EC 1.1.1.179,2DD, EC 1.3.1.20, Hum2DD, trans-1,2-dihydrobenzene-1,2-diol dehydrogenase | |
| Rabbit | |
| 36 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Expected to cross react based on sequence identity: Bovine (87%), Dog (84%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.99mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q9UQ10 | |
| DHDH | |
| Recombinant protein encompassing a sequence within the center region of human DHDH. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 27294 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction