missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHDH Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99195B
This item is not returnable.
View return policy
Description
DHDH Polyclonal antibody specifically detects DHDH in Human samples. It is validated for Western Blot
Specifications
| DHDH | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 27294 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Biotin | |
| 3-deoxyglucosone reductase, dihydrodiol dehydrogenase (dimeric), Dimeric dihydrodiol dehydrogenase, D-xylose 1-dehydrogenase, D-xylose-NADP dehydrogenase, EC 1.1.1.179,2DD, EC 1.3.1.20, Hum2DD, trans-1,2-dihydrobenzene-1,2-diol dehydrogenase | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human DHDH | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction