missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHDDS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15099-1-AP-150UL
This item is not returnable.
View return policy
Description
Dehydrodolichyl diphosphate (dedol-PP) synthase catalyzes cis-prenyl chain elongation to produce the polyprenyl backbone of dolichol, a glycosyl carrier lipid required for the biosynthesis of several classes of glycoproteins. Dehydrodolichyl diphosphate (dedol-PP) synthase catalyzes cis-prenyl chain elongation to produce the polyprenyl backbone of dolichol, a glycosyl carrier lipid required for the biosynthesis of several classes of glycoproteins.Specifications
| DHDDS | |
| Polyclonal | |
| Unconjugated | |
| DHDDS | |
| CIT, CPT, Dedol PP synthase, DHDDS, DS, HDS | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 298541, 67422, 79947 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.67 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q86SQ9, Q99KU1 | |
| Dhdds | |
| DHDDS Fusion Protein Ag7426 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction