missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DHDDS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555396
This item is not returnable.
View return policy
Description
Immunogen sequence: HSVLQKARDM YAEERKRQQL ERDQATVTEQ LLREGLQASG DAQLRRTRLH KLSARREERV QGFLQALELK RADWLARLGT ASA Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 89%.
Dehydrodolichyl diphosphate (dedol-PP) synthase catalyzes cis-prenyl chain elongation to produce the polyprenyl backbone of dolichol, a glycosyl carrier lipid required for the biosynthesis of several classes of glycoproteins. Dehydrodolichyl diphosphate (dedol-PP) synthase catalyzes cis-prenyl chain elongation to produce the polyprenyl backbone of dolichol, a glycosyl carrier lipid required for the biosynthesis of several classes of glycoproteins.
Specifications
| DHDDS | |
| Polyclonal | |
| Unconjugated | |
| DHDDS | |
| 3222401G21Rik; cis-IPTase; cis-isoprenyltransferase; cis-prenyl transferase; Cis-prenyltransferase subunit hCIT; CIT; CPT; dedol-PP synthase; dehydrodolichyl diphosphate syntase complex subunit Dhdds; dehydrodolichyl diphosphate synthase; dehydrodolichyl diphosphate synthase complex subunit DHDDS; dehydrodolichyl diphosphate synthase isoform a; dehydrodolichyl diphosphate synthase subunit; Dhdds; DS; epididymis tissue protein Li 189m; FLJ13102; HDS; RP59; W91638; wu:fd56c06; zgc:77088 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79947 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86SQ9 | |
| DHDDS | |
| Recombinant protein corresponding to Human DHDDS. Recombinant protein control fragment (Product #RP-94158). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction