missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DHCR7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560666
This item is not returnable.
View return policy
Description
Immunogen sequence: HQKDLFRRTD GRCLIWGRKP KVIECSYTSA DGQRHHSKLL VSGFWGVARH FNYVGDLMG Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 88%.
Production of cholesterol by reduction of C7-C8 double bond of 7-dehydrocholesterol (7-DHC).
Specifications
| DHCR7 | |
| Polyclonal | |
| Unconjugated | |
| DHCR7 | |
| 7-dehydrocholesterol reductase; 7-DHC reductase; AA409147; D7SR; delta-7-dehydrocholesterol reductase; Delta7-sterol reductase; Dhcr7; putative sterol reductase SR-2; SLOS; Sterol Delta(7)-reductase; sterol delta-7-reductase; Sterol reductase SR-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1717 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UBM7 | |
| DHCR7 | |
| Recombinant protein corresponding to Human DHCR7. Recombinant protein control fragment (Product #RP-91294). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction