missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DGUOK Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557051
This item is not returnable.
View return policy
Description
Immunogen sequence: STFVKLLTKT YPEWHVATEP VATWQNIQAA GTQKACTAQS LGNLLDMMYR EPARWSYTFQ TFSFLSRLKV QLEPFPEKLL QARKPVQI Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 77%.
Mitochondrial deoxyguanosine kinase (DGUOK) is required for the phosphorylation of several deoxyribonucleosides and certain purine deoxykribonucleoside analogs widely employed as antiviral and chemotherapeutic agents. Purine deoxyribonucleoside analogs are extensively used in treatment of lymphoproliferative disorders. These compounds are administered as pro-drugs, and their efficiency is dependent on intracellular phosphorylation to the corresponding triphosphates. In mammalian cells, the phosphorylation of purine deoxyribonucleosides is mediated predominantly by 2 deoxyribonucleoside kinases: cytosolic deoxycytidine kinase (DCK) and mitochondrial deoxyguanosine kinase (DGUOK also known as DGK). DGUOK expression is ubiquitous, with highest levels in muscle, brain, liver and lymphoid tissues. Defects in DGUOK are a cause of mitochondrial DNA depletion syndrome (MDS). MDS is a clinically heterogeneous group of disorders characterized by a reduction in mitochondrial DNA (mtDNA) copy number. Primary mtDNA depletion is inherited as an autosomal recessive trait and may affect single organs, typically muscle or liver, or multiple tissues.
Specifications
| DGUOK | |
| Polyclonal | |
| Unconjugated | |
| DGUOK | |
| deoxyguanosine kinase; deoxyguanosine kinase, mitochondrial; dGK; dGK1; dGK2; dGK3; DGUOK; MTDPS3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1716 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q16854 | |
| DGUOK | |
| Recombinant protein corresponding to Human DGUOK. Recombinant protein control fragment (Product #RP-95995). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction